Merck
  • Effect of vehicles and penetration enhancers on the in vitro percutaneous absorption of tenoxicam through hairless mouse skin.
Effect of vehicles and penetration enhancers on the in vitro percutaneous absorption of tenoxicam through hairless mouse skin.
International journal of pharmaceutics (2002-03-14)
Hye Sun Gwak, In Koo Chun
ABSTRACT
The effects of vehicles and penetration enhancers on the in vitro permeation of tenoxicam from saturated solutions through dorsal hairless mouse skin were investigated. Various types of vehicles, including ester-, alcohol-, and ether-types and their mixtures, were used as vehicles, and then a series of fatty acids and amines were employed as enhancers, respectively. Even though the fluxes of tenoxicam from saturated pure vehicles were generally low (0.1-1.1 microg/cm2 per h), the skin permeability of tenoxicam was significantly increased by the combination of diethylene glycol monoethyl ether (DGME) and propylene glycol monolaurate (PGML) or propylene glycol monocaprylate (PGMC); the highest fluxes were achieved at 40% of DGME in both of the two cosolvents. The marked synergistic enhancement was also obtained by using propylene glycol (PG)-oleyl alcohol (OAl) cosolvent. The greatest flux was attained by the addition of unsaturated fatty acids at 3% concentration to PG. But saturated fatty acids failed to show a significant enhancing effect. The enhancement factors with the addition of oleic acid (OA) or linoleic acid (LOA) to PG were 348 and 238, respectively. Tromethamine (TM) showed an enhancing effect by the increased solubility; however, triethanolamine (TEA) did not show a significant enhancing effect. Rather, it decreased the fluxes of tenoxicam when added to PG with fatty acids. The above results indicate that the combinations of lipophilic vehicles like OA, LOA or OAl and hydrophilic vehicles like PG can be used for enhancing the skin permeation of tenoxicam.
Sigma-Aldrich
Tenoxicam, NSAID
Pricing and availability is not currently available.