Merck
  • Direct-to-biology, automated, nano-scale synthesis, and phenotypic screening-enabled E3 ligase modulator discovery.
Direct-to-biology, automated, nano-scale synthesis, and phenotypic screening-enabled E3 ligase modulator discovery.
Nature communications (2023-12-20)
Zefeng Wang, Shabnam Shaabani, Xiang Gao, Yuen Lam Dora Ng, Valeriia Sapozhnikova, Philipp Mertins, Jan Krönke, Alexander Dömling
ABSTRACT
Thalidomide and its analogs are molecular glues (MGs) that lead to targeted ubiquitination and degradation of key cancer proteins via the cereblon (CRBN) E3 ligase. Here, we develop a direct-to-biology (D2B) approach for accelerated discovery of MGs. In this platform, automated, high throughput, and nano scale synthesis of hundreds of pomalidomide-based MGs was combined with rapid phenotypic screening, enabling an unprecedented fast identification of potent CRBN-acting MGs. The small molecules were further validated by degradation profiling and anti-cancer activity. This revealed E14 as a potent MG degrader targeting IKZF1/3, GSPT1 and 2 with profound effects on a panel of cancer cells. In a more generalized view, integration of automated, nanoscale synthesis with phenotypic assays has the potential to accelerate MGs discovery.
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)
Sigma-Aldrich
Lenalidomide, ≥98% (HPLC)
Sigma-Aldrich
QuicTPD Acid Optimization Set
Sigma-Aldrich
QuicTPD Amine Optimization Set
Sigma-Aldrich
QuicTPD Acid Screening Set
Sigma-Aldrich
QuicTPD Amine Screening Set