Merck
Enhanced uptake of [11C]TPMP in canine brain tumor: a PET study.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (1999-07-15)
I Madar, J H Anderson, Z Szabo, U Scheffel, P F Kao, H T Ravert, R F Dannals
ABSTRACT
In vitro studies have demonstrated the membrane potential-dependent enhanced uptake of phosphonium salts, including [3H]triphenylmethylphosphonium (TPMP), into mitochondria of carcinoma and glioma-derived tumor cells, suggesting the potential use of phosphonium salts as tracers for tumor imaging. This study characterizes the in vivo uptake of [11C]TPMP in canine brain glioma using PET. Dynamic paired PET studies of [11C]TPMP followed by [68Ga]ethylenediaminetetraacetic acid (EDTA) were performed 4 d before and 9 d after tumor cell inoculation. Graphical analysis was used to evaluate [11C]TPMP retention in tumor tissue. Distribution of tracer uptake was compared with tumor histological sections. [11C]TPMP exhibited enhanced uptake and prolonged retention in tumor cells. Patlak plot was linear over the 20- to 95-min postinjection period (r = 0.97 +/- 0.1). [68Ga]EDTA exhibited a gradual washout from the tumor tissue. The tumor-to-normal brain uptake ratio at 55 to 95 min postinjection was 47.5 for [11C]TPMP and 8.1 for [68Ga]EDTA. Qualitative comparison with histological sections indicated that [11C]TPMP enhanced uptake was restricted to the tumor area. The enhanced uptake and prolonged retention in tumor suggest [11C]TPMP as a promising means for imaging of gliomas in dogs. The need for studies in humans is indicated.
Sigma-Aldrich
Methyltriphenylphosphonium bromide, 98%