Merck
  • [Persistence properties of microflora of open water reservoirs and drinking water].
[Persistence properties of microflora of open water reservoirs and drinking water].
Zhurnal mikrobiologii, epidemiologii, i immunobiologii (2003-04-26)
I A Misetov, N V Nemtseva
ABSTRACT
As revealed in this study, the characteristics of the bacterial persistence of water microflora are suitable for the sanitary and hygienic characterization of natural water reservoirs and drinking water, as well as for evaluating the effectiveness of the antimicrobial action of disinfectants.
Sigma-Aldrich
N-Chlorobenzenesulfonamide sodium salt, ~28% active chlorine basis