Merck
  • Chemical modification of glucose oxidase: possible formation of molten globule-like intermediate structure.
Chemical modification of glucose oxidase: possible formation of molten globule-like intermediate structure.
FEBS letters (2004-03-12)
Saman Hosseinkhani, Bijan Ranjbar, Hossein Naderi-Manesh, Mohsen Nemat-Gorgani
ABSTRACT
Chemical modification of lysine residues in glucose oxidase was carried out using citraconic anhydride. Modification brought about changes in the kinetic properties of the enzyme as evident by substantial lowering of V(max) and K(m). Enhancement of tryptophan fluorescence was observed with a dramatic change in its pH dependence due to modification. Near- and far-UV circular dichroism spectra of the native and modified forms suggested formation of molten globule-like structures, further supported by 8-anilino-1-naphthalenesulfonic acid fluorescence which indicated higher exposure of hydrophobic residues as a result of chemical modification.
Sigma-Aldrich
Citraconic anhydride, 98%