Merck
  • Effects of polymorphisms in CYP2D6 and ABC transporters and side effects induced by gefitinib on the pharmacokinetics of the gefitinib metabolite, O-desmethyl gefitinib.
Effects of polymorphisms in CYP2D6 and ABC transporters and side effects induced by gefitinib on the pharmacokinetics of the gefitinib metabolite, O-desmethyl gefitinib.
Medical oncology (Northwood, London, England) (2016-05-08)
Hiroyuki Kobayashi, Kazuhiro Sato, Takenori Niioka, Masahide Takeda, Yuji Okuda, Mariko Asano, Hiroshi Ito, Masatomo Miura
ABSTRACT
We investigated the effects of polymorphisms in CYP2D6, ABCB1, and ABCG2 and the side effects induced by gefitinib on the pharmacokinetics of O-desmethyl gefitinib, the active metabolite of gefitinib. On day 14 after beginning therapy with gefitinib, plasma concentrations of gefitinib and O-desmethyl gefitinib were measured. Patients were grouped into three groups according to their combination of CYP2D6 alleles: homozygous extensive metabolisers (EMs; *1/*1, *1/*2, and *2/*2; n = 13), heterozygous EMs (*1/*5, *2/*5, *1/*10, and *2/*10; n = 18), and intermediate metabolisers (IMs; *5/*10 and *10/*10; n = 5). The median AUC0-24 of O-desmethyl gefitinib in CYP2D6 IMs was 1460 ng h/mL, whereas that in homozygous EMs was 12,523 ng h/mL (P = 0.021 in univariate analysis). The median AUC ratio of O-desmethyl gefitinib to gefitinib differed among homozygous EMs, heterozygous EMs, and IMs at a ratio of 1.41:0.86:0.24 (P = 0.030). On the other hand, there were no significant differences in the AUC0-24 of O-desmethyl gefitinib between ABCB1 and ABCG2 genotypes. In a multivariate analysis, CYP2D6 homozygous EMs (P = 0.012) were predictive for a higher AUC0-24 of O-desmethyl gefitinib. The side effects of diarrhoea, skin rash, and hepatotoxicity induced by gefitinib were unrelated to the AUC0-24 of O-desmethyl gefitinib. CYP2D6 polymorphisms were associated with the formation of O-desmethyl gefitinib from gefitinib. In CYP2D6 homozygous EMs, the plasma concentrations of O-desmethyl gefitinib were higher over 24 h after taking gefitinib than those of the parent compound; however, side effects induced by gefitinib were unrelated to O-desmethyl gefitinib exposure.
Sigma-Aldrich
O-Desmethyl gefitinib, ≥95%, research grade