Merck
  • Microtubule dynamics reconstituted in vitro and imaged by single-molecule fluorescence microscopy.
Microtubule dynamics reconstituted in vitro and imaged by single-molecule fluorescence microscopy.
Methods in cell biology (2010-05-15)
Christopher Gell, Volker Bormuth, Gary J Brouhard, Daniel N Cohen, Stefan Diez, Claire T Friel, Jonne Helenius, Bert Nitzsche, Heike Petzold, Jan Ribbe, Erik Schäffer, Jeffrey H Stear, Anastasiya Trushko, Vladimir Varga, Per O Widlund, Marija Zanic, Jonathon Howard
ABSTRACT
In vitro assays that reconstitute the dynamic behavior of microtubules provide insight into the roles of microtubule-associated proteins (MAPs) in regulating the growth, shrinkage, and catastrophe of microtubules. The use of total internal reflection fluorescence microscopy with fluorescently labeled tubulin and MAPs has allowed us to study microtubule dynamics at the resolution of single molecules. In this chapter we present a practical overview of how these assays are performed in our laboratory: fluorescent labeling methods, strategies to prolong the time to photo-bleaching, preparation of stabilized microtubules, flow-cells, microtubule immobilization, and finally an overview of the workflow that we follow when performing the experiments. At all stages, we focus on practical tips and highlight potential stumbling blocks.
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Pricing and availability is not currently available.