Merck
HomeBatteries, Supercapacitors & Fuel CellsRedi Dri™ Packaging for Consistent Free-Flow in Hygroscopic Salts
Redi Dri™ Packaging for Consistent Free-Flow in Hygroscopic Salts
Introduction
Hygroscopic, anhydrous inorganic salts are widely used in precision‑driven workflows, including pharmaceutical synthesis, semiconductor processing, and battery material preparation, where control of moisture during storage is relevant to material handling. Many inorganic salts readily absorb atmospheric moisture, leading to increased water content and, over time, clumping and reduced flowability. These changes can affect routine operations such as weighing, transfer, and dispensing under ambient storage conditions.
Maintaining controlled moisture levels during storage is essential to preserve the salts’ free-flowing properties, chemical integrity, and purity. Redi‑Dri™ packaging is designed to reduce moisture exposure during storage of hygroscopic salts used in moisture‑sensitive workflows.
Impact of Moisture on Hygroscopic Salt Flow
Moisture exposure can influence the handling and flow behavior of hygroscopic anhydrous salts used in moisture‑sensitive workflows. Even trace levels of water may promote hydrolysis in catalytic reactions or compromise precursor integrity during Chemical Vapor Deposition (CVD) and Atomic Layer Deposition (ALD). In applications such as catalytic processes and semiconductor deposition workflows, unintended moisture uptake may also contribute to variability in precursor behaviour during handling prior to use.
Many commonly used inorganic salts, including magnesium chloride and lithium chloride, readily absorb moisture during storage. Continued moisture uptake can increase interparticle interactions, leading to agglomeration, reduced flowability, and variability during weighing, transfer, and dispensing under ambient storage conditions.
Redi‑Dri™ packaging is designed to reduce moisture uptake in hygroscopic salts by providing controlled storage conditions within the container. Maintaining low moisture levels (below 50 ppm), as quantified by Karl Fischer titration, is associated with stable handling behavior during weighing, transfer, and dispensing under the evaluated conditions.
Time-Lapse Study: Moisture Absorption in Salts with Redi Dri™ Packaging
To assess moisture uptake over time, a controlled time‑lapse storage study was conducted in which moisture content was measured at predefined intervals using Karl Fischer (KF) titration. Samples packaged with Redi‑Dri™ sheets and samples in standard packaging were exposed to a controlled high‑humidity environment (>75 ± 5% relative humidity) to simulate challenging storage conditions.
Moisture content was monitored through monthly KF titration and visual inspection over a 12‑month period. Salts stored with Redi‑Dri™ packaging maintained low moisture levels and remained free‑flowing throughout the study duration, while samples stored in standard packaging exhibited progressive moisture uptake accompanied by visible agglomeration within two months.
Anhydrous lithium sulfate (99.5% trace metals basis) was evaluated as a representative hygroscopic salt under these conditions. Identical sample quantities were stored in standard and Redi‑Dri™ packaging and analysed at regular intervals, as shown in Figure 1.
Line chart showing Karl Fischer (KF) moisture content (%) of anhydrous lithium sulfate stored in Redi Dri™ packaging and in standard packaging at 0, 3, 6, and 12 months under high humidity conditions (>75 ± 5% RH).
Figure 1.Karl Fischer moisture content (%) of anhydrous lithium sulfate (99.5% trace metals basis) during 12 month storage under high humidity conditions (>75 ± 5% RH), comparing Redi Dri™ and standard packaging.
The Redi Dri™ sample maintains consistently low moisture content with minimal variation over time, while the control sample shows a steady increase in moisture content, consistent with progressive moisture uptake during storage.
Storage Stability Results Comparing Redi-Dri™ and Standard Packaging: The time lapse storage study revealed distinct differences in moisture uptake between hygroscopic salts stored in Redi Dri™ packaging and those stored in standard packaging. Over the 12 month study period, samples stored with Redi Dri™ packaging maintained low moisture content and remained free flowing, with no observable agglomeration.
In contrast, samples stored in standard packaging showed progressive moisture uptake accompanied by agglomeration within two months, resulting in reduced flowability and increased variability during routine handling operations such as weighing, transfer, and dispensing.
Quantitative Results
  • Sample with Redi Dri™  Packaging: Moisture content decreased from 0.20% to 0.17% over 12 months, indicating controlled moisture levels under the tested storage conditions.
  • Sample without Redi Dri packaging: Moisture content increased from 0.20% to 0.76% over the same period, consistent with progressive moisture uptake in unprotected hygroscopic inorganic salts.
These results demonstrate that traditional packaging conditions using Redi Dri™ support sustained low moisture content and stable handling properties for moisture sensitive inorganic salts used in energy storage and semiconductor related workflows.
ICP-MS/OES Verified Long-Term Trace Metals Purity
In applications involving hygroscopic inorganic salts, changes in trace metal impurity levels during storage may influence material consistency and reproducibility. Trace metal impurities are therefore monitored to ensure that storage conditions and packaging systems do not introduce contamination or alter impurity profiles over time. Analytical techniques such as Inductively Coupled Plasma Mass Spectrometry (ICP-MS) and Inductively Coupled Plasma Optical Emission Spectroscopy (ICP-OES) are commonly used for this purpose due to their sensitivity and multi element capability.
To evaluate trace metal impurity stability during storage, samples packaged with Redi Dri™ sheets and samples stored in standard packaging were analysed at predefined intervals, including Day 0 (baseline), Day 90, Day 180, Day 270, and Day 365. Samples were stored under controlled conditions representative of typical storage environments.
Across the monitored storage period, ICP-MS analysis showed no increases in trace metal impurity levels consistent with leaching, adsorption, or other interactions between the packaging components and the stored material. As shown in Figure 2, ICP-MS analysis of anhydrous lithium sulfate (99.5% trace metals basis; Trace Metal Analysis < 5000 ppm) was performed to monitor trace metal impurities during storage over a 12 month period.
The data compare samples stored with Redi Dri™ packaging to control samples stored without the Redi Dri™ sheet. Analysis of a 32 element panel showed less than 2% variation in measured impurity levels over time, indicating stable trace metal profiles throughout the study duration under the evaluated storage conditions.
Multi line chart showing concentrations of selected trace metals (including sodium, calcium, zinc, and total trace metals) measured by ICP MS in anhydrous lithium sulfate at baseline, 3 months, 6 months, 10 months, and 12 months for samples stored in Redi Dri™ packaging and standard packaging.
Figure 2.Trace metal impurity levels measured by ICP MS for anhydrous lithium sulfate (99.5% trace metals basis; < 5000 ppm) during 12 month storage, comparing Redi Dri™ and standard packaging.
Assay Stability
Assay values were monitored over the storage period to evaluate whether prolonged moisture exposure and storage conditions led to measurable chemical degradation or impurity formation. Assay stability serves as an indicator of chemical integrity, particularly for hygroscopic inorganic salts that may be susceptible to moisture induced reactions during storage.
As shown in Figure 3, assay measurements for anhydrous lithium sulfate (99.5% trace metals basis) remained stable over the 12 month storage period for samples stored with Redi Dri™ packaging. No systematic decrease in assay values was observed when compared to baseline measurements, indicating the absence of measurable degradation under the tested conditions.
These observations suggest that controlled storage conditions maintained using Redi Dri™ packaging support preservation of chemical assay values during extended storage, consistent with maintained material integrity for hygroscopic inorganic salts.
Line chart showing assay percentage of anhydrous lithium sulfate (99.5% trace metals basis) measured at 0, 3, 6, and 12 months for samples stored in Redi Dri™ packaging and standard packaging.
Figure 3.Assay values (%) of anhydrous lithium sulfate (99.5% trace metals basis) measured over 12 month storage, comparing Redi Dri™ and standard packaging.
Conclusion
Hygroscopic anhydrous inorganic salts are susceptible to moisture uptake during storage, which can lead to changes in flow behavior, handling variability, and, in some cases, chemical or impurity related concerns. Over time, these effects may influence material consistency and reproducibility in applications such as pharmaceutical synthesis, semiconductor processing, and battery material preparation.The findings below summarize the impact of controlled moisture conditions on the stability and handling behaviour of inorganic salts stored with Redi Dri™ packaging.
Key Findings: Using Redi Dri™ Packaging
  • Moisture control: Karl Fischer titration performed under high humidity conditions (>75 ± 5% RH) showed that salts stored with Redi Dri™ packaging maintained low moisture content over the study period, with a measurable reduction in moisture content observed over time.
  • Assay stability: Assay values remained stable over 12 months (Figure 3), with no measurable evidence of chemical degradation, hydrolysis, or impurity formation under the tested conditions.
  • Trace Metals Stability: ICP MS analysis across a 32 element panel showed less than 2% variation over 12 months, consistent with stable trace metal impurity profiles and no evidence of leaching or interaction between the packaging components and the stored material.
  • Visual Free-Flow Reliability: Materials stored with Redi Dri™ packaging remained free flowing and pourable throughout the 12 month storage period, with no visible agglomeration observed.
Overall, the observations indicate that controlled moisture conditions provided by Redi Dri™ packaging support stable handling characteristics, preserved chemical integrity, and consistent trace metal impurity profiles in hygroscopic inorganic salts during storage. The combination of low moisture content, stable assay values, and unchanged trace metal levels corresponds to reduced clumping risk and maintained material quality over time, with relevance for moisture sensitive storage workflows used in pharmaceutical synthesis, semiconductor CVD/ALD precursor handling, and battery electrolyte preparation.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500