Iupac Name
5-[4-fluoro-3-(trifluoromethyl)phenyl]-2-methoxypyridine
Inchi Code
1S/C13H9F4NO/c1-19-12-5-3-9(7-18-12)8-2-4-11(14)10(6-8)13(15,16)17/h2-7H,1H3
InChI key
FHIODCIAGGZQPW-UHFFFAOYSA-N
Purity
97%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0635
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?LD-0635
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service