- Polymeric microspheres with N-methyl-d-glucamine ligands for boron removal from water solution by adsorption?membrane filtration process.
Polymeric microspheres with N-methyl-d-glucamine ligands for boron removal from water solution by adsorption?membrane filtration process.
Environmental Geochemistry and Health (2010)
Wolska J