Merck
  • Potential role of CBX7 in regulating pluripotency of adult human pluripotent-like olfactory stem cells in stroke model.
Potential role of CBX7 in regulating pluripotency of adult human pluripotent-like olfactory stem cells in stroke model.
Cell death & disease (2018-05-03)
Jia-Rong Fan, Hsu-Tung Lee, Wei Lee, Chen-Huan Lin, Chun Y Hsu, Chia-Hung Hsieh, Woei-Cherng Shyu
ABSTRACT
The adult olfactory mucosa, a highly regenerative tissue with unique life-long neurogenesis ability, is thought to harbor a naïve yet tightly controlled stem cell population. It will provide unique benefits in various stem cell-based therapies, such as stroke treatment. Here, we identified a subpopulation of adult pluripotent-like olfactory stem cells (APOSCs), which were modulated by an epigenetic repressor of CBX7. APOSCs form a floating sphere, express pluripotency markers Nanog, Oct-4, Sox-2, and SSEA-4 and show alkaline phosphatase activity. In addition, APOSCs display self-renewal and a pluripotent potential to differentiate into all three germ layers. Moreover, APOSCs coexpress pluripotency markers with CBX7. Within their natural niche, APOSCs from CBX7+/+ mice responded promptly to either spontaneous or injury-induced tissue regeneration. However, APOSCs from CBX7-/- mice manifested an impaired self-renewal and differentiation potential. Similarly, in vitro-cultivated CBX7-/- APOSCs underwent premature senescence, whereas CBX7+/+ APOSCs still actively divided, indicating that CBX7 is required for the self-renewal of APOSCs. Intracerebral implantation of APOSCs improved the stroke-mediated neurological dysfunction in rodents. These findings indicate that CBX7 plays a critical role in the regenerative properties of APOSCs and indicate the safety and feasibility of implantation of autologous APOSCs in stroke treatment.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture