Merck
  • Nutritive value of multienzyme supplemented cold-pressed camelina cake for pigs.
Nutritive value of multienzyme supplemented cold-pressed camelina cake for pigs.
Journal of animal science (2018-02-01)
T A Woyengo, R Patterson, C L Levesque
ABSTRACT
Cold-pressed camelina cake (CPCC) is a fibrous co-product of camelina seed pressing and available for livestock feeding. However, information is lacking on the effect of supplementing fiber-degrading enzymes to CPCC-based diets on nutrient utilization by pigs. Experiment 1 determined the effect of multienzyme supplementation on standardized ileal digestibility (SID) of amino acid (AA) and net energy (NE) value of CPCC for pigs. Six ileal-cannulated barrows (average initial body weight [BW] = 36 kg) were fed five diets in 5 × 5 Latin square design with 1 added column to give six replicates per diet. The diets were a corn-soybean meal (SBM)-soybean oil-based diet and the basal diet with corn, SBM, and soybean oil replaced by 25% CPCC with or without multienzyme (600 U of xylanase, 75 U of glucanase, 250 U of cellulose, 30 U of mannanase, 350 U of invertase, 2,500 U of protease, and 6,000 U of amylase/kg of diet; Superzyme-CS, 0.5 g/kg) in a 2 × 2 factorial arrangement. The fifth diet was a low-casein cornstarch-based diet. The ratio of corn to SBM and soybean oil in the basal diet was identical to the CCPC-containing diets to allow calculation of nutrient digestibility of CPCC by the difference method. On a dry matter (DM) basis, CPCC contained 42% crude protein, 10.5% ether extract, 25.4% neutral detergent fiber (NDF), 2.07% Lys, 0.73% Met, 1.64% Thr, 0.51% Trp, and 22.1 trypsin inhibitor units per milligram, respectively. The SID of Lys, Met, Thr, and Trp for CPCC were 43.5%, 70.7%, 44.8%, and 55.3%, respectively. The digestible energy (DE) and NE values for CPCC were 3,663 and 2,209 kcal/kg of DM, respectively. Multienzyme supplementation did not affect the SID of AA, and DE and NE values for the corn-SBM-CPCC-based diet, and for the CPCC. In experiment 2, the effects of multienzyme dosage (0.5 or 50 g/kg of treated feedstuff) on porcine in vitro digestibility of DM (IVDDM) of CPCC was determined. The IVDDM of CPCC was increased (P < 0.001) with an increase in multienzyme dosage. Multienzyme at 0.5 g/kg did not affect IVDDM of CPCC. However, multienzyme at 50 g/kg increased (P < 0.01) IVDDM for CPCC by at least 16%. In conclusion, multienzyme at 0.5 g/kg did not affect SID of AA and NE values, and IVDDM for CPCC. However, multienzyme at 50 g/kg improved IVDDM of CPCC, implying that the efficacy of the multienzyme with regard to improving nutrient digestibility of CPCC in pigs is dosage dependent.
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications