Merck
  • Enantioselective hydrogenation of acyclic aromatic N-aryl imines catalyzed by an iridium complex of (S, S)-1, 2-bis (tert-butylmethylphosphino) ethane.
Enantioselective hydrogenation of acyclic aromatic N-aryl imines catalyzed by an iridium complex of (S, S)-1, 2-bis (tert-butylmethylphosphino) ethane.
Organic Letters (2006)
Imamoto T, et al.