Merck
  • Association of the-844G> A polymorphism in the catalase gene with the increased risk of essential hypertension in smokers.
Association of the-844G> A polymorphism in the catalase gene with the increased risk of essential hypertension in smokers.
Terapevticheski? Arkhiv (2016)
Bushueva O Y, et al.