Merck
  • Determination of volatile compounds by GC-IMS to assign the quality of virgin olive oil
Determination of volatile compounds by GC-IMS to assign the quality of virgin olive oil
Food Chemistry (2015)
Garrido-Delgado, Rocio and del Mar Dobao-Prieto, Maria and Arce, Lourdes and Valcarcel, Miguel