Merck
  • Anillin is required for tumor growth and regulated by miR-15a/miR-16-1 in HBV-related hepatocellular carcinoma.
Anillin is required for tumor growth and regulated by miR-15a/miR-16-1 in HBV-related hepatocellular carcinoma.
Aging (2018-08-14)
Yi-Fan Lian, Yan-Lin Huang, Jia-Liang Wang, Mei-Hai Deng, Tian-Liang Xia, Mu-Sheng Zeng, Min-Shan Chen, Hong-Bo Wang, Yue-Hua Huang
ABSTRACT
Anillin (ANLN) is an actin-binding protein essential for assembly of cleavage furrow during cytokinesis. Although reportedly overexpressed in various human cancers, its role in hepatocellular carcinoma (HCC) is unclear. To address this issue, we confirmed that in 436 liver samples obtained from surgically removed HCC tissues, higher ANLN expression was detected in tumor tissues than in adjacent non-tumor tissues of HCC as measured by immunohistochemistry, quantitative real-time PCR and western blotting. Correlation and Kaplan-Meier analysis revealed that patients with higher ANLN expression were associated with worse clinical outcomes and a shorter survival time, respectively. Moreover, ANLN inhibition resulted in growth restraint, reduced colony formation, and a lower sphere number in suspension culture. Mechanistically, ANLN deficiency induced an increasing number of multinucleated cells along with the activation of apoptosis signaling and DNA damage checkpoints. Furthermore, HBV infection increased ANLN expression by inhibiting the expression of microRNA (miR)-15a and miR-16-1, both of which were identified as ANLN upstream repressors by targeting its 3' untranslated region. Thus, we conclude that ANLN promotes tumor growth by ways of decreased apoptosis and DNA damage. Expression level of ANLN significantly influences the survival probability of HCC patients and may represent a promising prognostic biomarker.
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis