Merck
The acylation of lipophilic alcohols by lysosomal phospholipase A2.
Journal of lipid research (2007-07-14)
Akira Abe, Miki Hiraoka, James A Shayman
ABSTRACT
A novel lysosomal phospholipase A(2) (LPLA2) with specificity toward phosphatidylethanolamine and phosphatidylcholine was previously purified and cloned. LPLA2 transfers sn-1 or sn-2 acyl groups of phospholipids to the C1 hydroxyl of the short-chain ceramide N-acetylsphingosine (NAS) under acidic conditions. The common features of lipophilic alcohols serving as acceptor molecules in the transacylase reaction were examined. 1-O-Hexadecyl-2-acetyl-sn-glycerol (HAG) was acylated by LPLA2 similar to NAS. HAG competed with NAS and inhibited NAS acylation. The transacylation of 1-O-hexadecyl-glycerol (HG), 1-O-palmityl-2-O-methyl-sn-glycerol (PMG), and monoacylglycerols was also investigated. HG, PMG, 1- or 3-palmitoyl-sn-glycerol, and 2-palmitoylglycerol were converted to 1,3-alkylacylglycerol, 1,2-dialkyl-3-acylglycerol, 1,3-diacylglycerol, and 1,2- or 2,3-diacylglycerol, respectively. HG and monoacylglycerol inhibited the acylation of NAS by the enzyme with IC(50) values of 35 and 45 microM, respectively. Additionally, the enzyme acylated glycerol to produce 1- or 3-acyl-sn-glycerol but not 2-acylglycerol. Therefore, the preferred acceptor molecules for LPLA2 are primary alcohols with one long carbon chain and one small nonpolar residue linked to the C2 position of ethanol. The enzyme acylated other natural lipophilic alcohols, including anandamide and oleoylethanolamide. Thus, LPLA2 may function to remodel acyl groups and modulate the biological and pharmacological activities of some lipophilic alcohols.
Product Description
Avanti
1-C16 Ether MG, Avanti Research - A Croda Brand 999971P, powder