Merck
  • Geminin is an indispensable inhibitor of Cdt1 in mouse embryonic stem cells.
Geminin is an indispensable inhibitor of Cdt1 in mouse embryonic stem cells.
Genes to cells : devoted to molecular & cellular mechanisms (2017-03-16)
Masaki Hosogane, Lena Bosu, Emiko Fukumoto, Hidetoshi Yamada, Soichiro Sato, Keiko Nakayama
ABSTRACT
Geminin is implicated in regulation of the cell cycle and differentiation. Although loss of Geminin triggers unscheduled DNA rereplication as a result of interruption of its interaction with Cdt1 in some somatic cancer cells, whether such cell cycle regulation also operates in embryonic stem cells (ESCs) has remained unclear. To characterize the Geminin-Cdt1 axis in ESCs and compare it with that in somatic cells, we established conditional knockout (KO) of Geminin in mouse ESCs and mouse embryonic fibroblasts (MEFs). Geminin KO ESCs manifest a large flattened morphology, develop polyploidy accompanied by DNA damage and G2 -M checkpoint activation, and subsequently undergo apoptosis. Rereplication in Geminin KO ESCs was attenuated by inhibition of G2 -M checkpoint signaling or by expression of wild-type Geminin, but not by expression of a Geminin mutant that does not bind to Cdt1, indicating the importance of sequestration of Cdt1 by Geminin in G2 phase. In contrast, Geminin KO MEFs did not manifest disturbance of the cell cycle unless they were treated to force abnormal accumulation of Cdt1. Together, our results indicate that Geminin is a key inhibitor of Cdt1 in mouse ESCs, but that it plays a backup role in MEFs to compensate for accidental up-regulation of Cdt1.
Sigma-Aldrich
Anti-phospho-H2A.X (Ser139) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit