Merck
  • FGF-induced Pea3 transcription factors program the genetic landscape for cell fate determination.
FGF-induced Pea3 transcription factors program the genetic landscape for cell fate determination.
PLoS genetics (2018-09-07)
Ankur Garg, Abdul Hannan, Qian Wang, Tamica Collins, Siying Teng, Mukesh Bansal, Jian Zhong, Keli Xu, Xin Zhang
ABSTRACT
FGF signaling is a potent inducer of lacrimal gland development in the eye, capable of transforming the corneal epithelium into glandular tissues. Here, we show that genetic ablation of the Pea3 family of transcription factors not only disrupted the ductal elongation and branching of the lacrimal gland, but also biased the lacrimal gland epithelium toward an epidermal cell fate. Analysis of high-throughput gene expression and chromatin immunoprecipitation data revealed that the Pea3 genes directly control both the positive and negative feedback loops of FGF signaling. Importantly, Pea3 genes are also required to suppress aberrant Notch signaling which, if gone unchecked, can compromise lacrimal gland development by preventing the expression of both Sox and Six family genes. These results demonstrate that Pea3 genes are key FGF early response transcriptional factors, programing the genetic landscape for cell fate determination.