Merck
  • Down-regulation of miR-327 alleviates ischemia/reperfusion-induced myocardial damage by targeting RP105
Down-regulation of miR-327 alleviates ischemia/reperfusion-induced myocardial damage by targeting RP105
Cellular Physiology and Biochemistry (2018)
Yang Y, et al.