Merck
  • Valsartan, captopril, or both in myocardial infarction complicated by heart failure, left ventricular dysfunction, or both
Valsartan, captopril, or both in myocardial infarction complicated by heart failure, left ventricular dysfunction, or both
The New England Journal of Medicine (2003)
Pfeffer MA, et al.