Merck
  • A metabolic pathway assembled by enzyme selection may support herbivory of leaf-cutter ants on plant starch
A metabolic pathway assembled by enzyme selection may support herbivory of leaf-cutter ants on plant starch
Journal of Insect Physiology (2013)
Bacci Jr M, et al.