Merck
  • Omega-6 vegetable oils as a driver of coronary heart disease: the oxidized linoleic acid hypothesis
Omega-6 vegetable oils as a driver of coronary heart disease: the oxidized linoleic acid hypothesis
Open heart (2018)
DiNicolantonio JJ and O?Keefe JH