Merck
  • Combination therapy targeting the tumor microenvironment is effective in a model of human ocular melanoma
Combination therapy targeting the tumor microenvironment is effective in a model of human ocular melanoma
Journal of Translational Medicine (2007)
Mangiameli DP, et al.