Merck
  • The Influence of TGF-β3, EGF, and BGN on SOX9 and RUNX2 Expression in Human Chondrogenic Progenitor Cells.
The Influence of TGF-β3, EGF, and BGN on SOX9 and RUNX2 Expression in Human Chondrogenic Progenitor Cells.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2018-11-16)
Jerome Nicolas Janssen, Sarah Batschkus, Stefan Schimmel, Christa Bode, Boris Schminke, Nicolai Miosge
ABSTRACT
Osteoarthritis (OA) is the most common chronic joint disease and leads to the degradation of the extracellular matrix by an imbalance between anabolic and catabolic processes. TGF-β3 (transforming growth factor beta-3) and epidermal growth factor (EGF) influence the osteochondrogenic potential of chondrocytes. In this study, we compared the expression of mediators and receptors in the TGF-β3 and EGF pathways, as well as biglycan (BGN), in healthy and diseased chondrocytes. Furthermore, we used chondrogenic progenitor cells (CPCs) for in vitro stimulation and knockdown experiments to elucidate the effects of TGF-β3 and EGF on the chondrogenic potential. Our results demonstrate that the expression of TGF-beta receptor type-1 (TGFBRI) and epidermal growth factor receptor (EGFR) is altered in diseased chondrocytes as well as in CPCs. Moreover, TGF-β3 and EGF stimulation influenced the expression levels of BGN, SRY (sex determining region Y)-box 9 (SOX9), and Runt-related transcription factor 2 (RUNX2) in CPCs. Therefore, changes in TGFBRI and EGFR expression likely contribute to the degenerative and regenerative effects seen in late stages of OA.