Merck
  • Analysis of captopril and hydrochlorothiazide combination tablet formulations by liquid chromatography
Analysis of captopril and hydrochlorothiazide combination tablet formulations by liquid chromatography
Journal of Pharmaceutical Sciences (1984)
Kirschbaum J and Perlman S