Merck
  • Fas gene polymorphisms in systemic lupus erythematosus and serum levels of some apoptosis-related molecules
Fas gene polymorphisms in systemic lupus erythematosus and serum levels of some apoptosis-related molecules
Immunological Investigations (2010)
Araste JM, et al.