Merck
  • Quantitative determination of hemoglobin and cytochemical staining for peroxidase using 3, 3?, 5, 5?-tetramethylbenzidine dihydrochloride, a safe substitute for benzidine
Quantitative determination of hemoglobin and cytochemical staining for peroxidase using 3, 3?, 5, 5?-tetramethylbenzidine dihydrochloride, a safe substitute for benzidine
Analytical biochemistry (1979)
Liem HH, et al.