Merck
  • Specificity and kinetics of interstrand and intrastrand bifunctional alkylation by nitrogen mustards at a G-G-C sequence.
Specificity and kinetics of interstrand and intrastrand bifunctional alkylation by nitrogen mustards at a G-G-C sequence.
Nucleic acids research (1997-03-15)
G B Bauer, L F Povirk
ABSTRACT
Previous work showed that melphalan-induced mutations in the aprt gene of CHO cells are primarily transversions and occur preferentially at G-G-C sequences, which are potential sites for various bifunctional alkylations involving guanine N-7. To identify the DNA lesion(s) which may be responsible for these mutations, an end-labeled DNA duplex containing a frequent site of melphalan-induced mutation in the aprt gene was treated with melphalan, mechlorethamine or phosphoramide mustard. The sequence specificity and kinetics of formation of both interstrand and intrastrand crosslinks were determined. All mustards selectively formed two base-staggered interstrand crosslinks between the 5'G and the G opposite C in the 5'G-G-C sequence. Secondary alkylation was much slower for melphalan than for the other mustards and the resulting crosslink was more stable. Mechlorethamine and phosphoramide mustard induced intrastrand crosslinks between the two contiguous Gs in the G-G-C sequence in double-stranded DNA, but melphalan did not. Molecular dynamic simulations provided a structural explanation for this difference, in that the monofunctionally bound intermediates of mechlorethamine and phosphoramide mustard assumed thermodynamically stable conformations with the second arm in a position appropriate for intrastrand crosslink formation, while the corresponding melphalan monoadduct did not.