- Supramolecular architecture of new lanthanide coordination polymers of 2-aminoterephthalic acid and 1, 10-phenanthroline
Supramolecular architecture of new lanthanide coordination polymers of 2-aminoterephthalic acid and 1, 10-phenanthroline
New. J. Chem. (2004)
Liu CB, et al.