- An unexpected bifunctional acid base catalysis in IRMOF-3 for Knoevenagel condensation reactions
An unexpected bifunctional acid base catalysis in IRMOF-3 for Knoevenagel condensation reactions
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association (2012)
Llabres i Xamena FX, et al.