Merck
  • Affinity pulldown of γ-secretase and associated proteins from human and rat brain.
Affinity pulldown of γ-secretase and associated proteins from human and rat brain.
Journal of cellular and molecular medicine (2009-09-17)
Yasuhiro Teranishi, Ji-Yeun Hur, Hedvig Welander, Jenny Frånberg, Mikio Aoki, Bengt Winblad, Susanne Frykman, Lars O Tjernberg
ABSTRACT
γ-Secretase is a transmembrane protease complex responsible for the processing of a multitude of type 1 transmembrane proteins, including amyloid precursor protein (APP) and Notch. A functional complex is dependent on the assembly of four proteins: presenilin (PS), nicastrin, Aph-1 and Pen-2. Little is known about how the substrates are selected by γ-secretase, but it has been suggested that γ-secretase associated proteins (GSAPs) could be of importance. For instance, it was recently reported from studies in cell lines that TMP21, a transmembrane protein involved in trafficking, binds to γ-secretase and regulates the processing of APP-derived substrates without affecting Notch cleavage. Here, we present an efficient and selective method for purification and analysis of γ-secretase and GSAPs. Microsomal membranes were prepared from rat or human brain and incubated with a γ-secretase inhibitor coupled to biotin via a long linker and a S-S bridge. After pulldown using streptavidin beads, bound proteins were eluted under reducing conditions and digested by trypsin. The tryptic peptides were subjected to LC-MS/MS analysis, and proteins were identified by sequence data from MS/MS spectra. All of the known γ-secretase components were identified. Interestingly, TMP21 and the PS associated protein syntaxin1 were associated to γ-secretase in rat brain. We suggest that the present method can be used for further studies on the composition of the γ-secretase complex.
Sigma-Aldrich
Anti-Presenilin-1 Antibody, loop, a.a. 263-378, CT, clone PS1-loop, ascites fluid, clone PS1-loop, Chemicon®