Merck
  • Safe and Efficacious Diphtheria Toxin-Based Treatment for Melanoma: Combination of a Light-On Gene-Expression System and Nanotechnology.
Safe and Efficacious Diphtheria Toxin-Based Treatment for Melanoma: Combination of a Light-On Gene-Expression System and Nanotechnology.
Molecular pharmaceutics (2019-11-26)
Jiajun Xu, Muye He, Xinyu Hou, Yan Wang, Chenting Shou, Xiaoran Cai, Zeting Yuan, Yu Yin, Minbo Lan, Kaiyan Lou, Yuzheng Zhao, Yi Yang, Xianjun Chen, Feng Gao
ABSTRACT
The controversy surrounding the use of diphtheria toxin (DT) as a therapeutic agent against tumor cells arises mainly from its unexpected harmfulness to healthy tissues. We encoded the cytotoxic fragment A of DT (DTA) as an objective gene in the Light-On gene-expression system to construct plasmids pGAVPO (pG) and pU5-DTA (pDTA). Meanwhile, a cRGD-modified ternary complex comprising plasmids, chitosan, and liposome (pG&pDTA@cRGD-CL) was prepared as a nanocarrier to ensure transfection efficiency. Benefiting from spatiotemporal control of this light-switchable transgene system and the superior tumor targeting of the carrier, toxins were designed to be expressed selectively in illuminated lesions. In vitro studies suggested that pG&pDTA@cRGD-CL exerted arrest of the S phase in B16F10 cells upon blue light irradiation and, ultimately, induced the apoptosis and necrosis of tumor cells. Such DTA-based treatment exerted enhanced antitumor activity in mice bearing B16F10 xenografts and displayed prolonged survival time with minimal side effects. Hence, we described novel DTA-based therapy combined with nanotechnology and the Light-On gene-expression system: such treatment could be a promising strategy against melanoma.
Product Description
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand