Merck
  • The effect of human interferon alpha on replication of different bovine viral diarrhea virus strains.
The effect of human interferon alpha on replication of different bovine viral diarrhea virus strains.
Acta virologica (2019-09-12)
A A Elsheikh, L J Braun, S M G Mansour, A Orabi, A S Alqahtani, D A Benfield, C C L Chase
ABSTRACT
Bovine viral diarrhea virus (BVDV) exists in two main biotypes: cytopathic (cp) and noncytopathic (ncp). Although some studies were done on the effect of interferon alpha (IFN-α) on BVDV, the effect of exogenous IFN against BVDV biotypes remains unclear. In the present study, we evaluated the comparative effect of exogenous human IFN-α (HuIFN-α) on different BVDV biotypes and genotypes. The results showed that exogenous HuIFN-α greatly inhibited the growth of different BVDV biotypes and genotypes. However, HuINF-α has a significant inhibitory effect on cp biotype compared to ncp one without significant variation between different genotypes. The effect of HuIFN-α on BVDV reached the maximum level at early stages of infection (0-20 h post infection) and increased in a dose-dependent manner (10-500 U/ml). Quantitative real-time RT-PCR was used to evaluate the effect of exogenous HuIFN-α on RNA synthesis of both BVDV biotypes. HuIFN-α reduced RNA production of cp by 4 logs compared to only 2 logs for ncp strains. Additionally, the antiviral effect of IFN-α against both BVDV biotypes seems to be independent of the RNA-dependent protein kinase (PKR) activation as assayed by direct analysis of in vivo phosphorylation of eIF2-α and by 2-aminopurine (2-AP) treatment. Collectively, these results indicated that the exogenous HuIFN-α treatment has an inhibitory effect not only on cp BVDV biotype but also on the ncp BVDV. The antiviral effect of exogenous HuIFN-α was biotype, time, dose but not genotype dependent. PKR has no role in the inhibitory effect suggesting that other IFN-antiviral pathways were involved. Keywords: BVDV biotypes; HuIFN-α; RNA synthesis; PKR-independent.
Product Description
Mepyramine impurity C, European Pharmacopoeia (EP) Reference Standard