Merck
  • Effects of pre-starter and starter diets varying in amino acid density given to broiler chickens that received coccidiosis vaccination at hatch1.
Effects of pre-starter and starter diets varying in amino acid density given to broiler chickens that received coccidiosis vaccination at hatch1.
Poultry science (2019-05-08)
S E Cloft, S J Rochell, K S Macklin, W A Dozier
ABSTRACT
An experiment was conducted to evaluate broilers vaccinated against coccidiosis fed combinations of pre-starter and starter diets varying in digestible amino acid (dAA) density from 1 to 19 d of age on subsequent growth performance and meat yield. A total of 1,800 Yield Plus Ross × Ross 708 male broilers were allocated to 60 floor pens and assigned to 1 of the 6 treatments. Four pre-starter diets varying in dAA density [1.15% dLys, 1.25% dLys, 1.35% dLys, and Positive Control (1.15% dLys + Diclazuril)] were fed until 9 d of age. Then, 3 starter diets varying in dAA density [1.15% dLys, 1.25% dLys, and PC (1.15% dLys + Diclazuril)] were given from 10 to 19 d of age. All diets were formulated to similar dAA ratios to dLys; hence, resulting in increasing amino acid (AA) density among the dietary treatments. Birds were given common grower and finisher diets for the remainder of the experiment. At 19, 27, and 40 d of age, broilers that received the 1.35% and 1.25% dLys density pre-starter and 1.25% dLys density starter diets had increased BW gain (P ≤ 0.006) compared with broilers fed the 1.15% dLys density diets. Broilers that received the 1.15% dLys density pre-starter diet had a lower cumulative BW gain (P = 0.007) than those fed the 1.35% and 1.25% dLys density pre-starter diets. Broilers that received the 1.35% and 1.25% dLys density pre-starter and 1.25% dLys density starter diets had the heaviest (P ≤ 0.008) total breast weights. Increasing the pre-starter dAA density enhanced (P ≤ 0.03) total breast weight by 28 g (1.25% dLys density) and 51 g (1.35% dLys density). Results from this study indicated that increasing dAA density from 1 to 19 d of age enabled a 1 to 19 d dAA intake increase of 4.8 g, which enhanced cumulative growth and meat yield of broilers vaccinated against coccidiosis.