Merck
  • Magnetite nanoparticles stabilized with polymeric bilayer of poly (ethylene glycol) methyl ether-poly (?-caprolactone) copolymers
Magnetite nanoparticles stabilized with polymeric bilayer of poly (ethylene glycol) methyl ether-poly (?-caprolactone) copolymers
Polymer (2008)
Meerod S, et al.