Merck
  • Targeted deletion of Insm2 in mice result in reduced insulin secretion and glucose intolerance.
Targeted deletion of Insm2 in mice result in reduced insulin secretion and glucose intolerance.
Journal of translational medicine (2018-10-26)
Lin Wang, Zhong Sheng Sun, Bingwu Xiang, Chi-Ju Wei, Yan Wang, Kevin Sun, Guanjie Chen, Michael S Lan, Gilberto N Carmona, Abner L Notkins, Tao Cai
ABSTRACT
Neurogenin3 (Ngn3) and neurogenic differentiation 1 (NeuroD1), two crucial transcriptional factors involved in human diabetes (OMIM: 601724) and islet development, have been previously found to directly target to the E-boxes of the insulinoma-associated 2 (Insm2) gene promoter, thereby activating the expression of Insm2 in insulin-secretion cells. However, little is known about the function of Insm2 in pancreatic islets and glucose metabolisms. Homozygous Insm2-/- mice were generated by using the CRISPR-Cas9 method. Glucose-stimulated insulin secretion and islet morphology were analyzed by ELISA and immunostainings. Expression levels of Insm2-associated molecules were measured using quantitative RT-PCR and Western blots. Fasting blood glucose levels of Insm2-/- mice were higher than wild-type counterparts. Insm2-/- mice also showed reduction in glucose tolerance and insulin/C-peptide levels when compared to the wild-type mice. RT-PCR and Western blot analysis revealed that expression of Insm1 was significantly increased in Insm2-/- mice, suggesting a compensatory response of the homolog gene Insm1. Similarly, transcriptional levels of Ngn3 and NeuroD1 were also increased in Insm2-/- mice. Moreover, Insm2-/- female mice showed a significantly decreased reproductive capacity. Our findings suggest that Insm2 is important in glucose-stimulated insulin secretion and is involved in the development pathway of neuroendocrine tissues which are regulated by the transcription factors Ngn3, NeuroD1 and Insm1.