Merck
  • Adhesion-independent alpha6beta4 integrin clustering is mediated by phosphatidylinositol 3-kinase.
Adhesion-independent alpha6beta4 integrin clustering is mediated by phosphatidylinositol 3-kinase.
Cancer research (2004-10-20)
Michael Z Gilcrease, Xiao Zhou, Kristin Welch
ABSTRACT
Clustering of cell-surface integrins is known to augment integrin-mediated signal transduction, but mechanisms of integrin clustering are poorly understood. Here we report that adhesion-independent clustering of alpha6beta4 integrin, known to be important in mediating tumor cell motility, is driven by phosphatidylinositol 3-kinase (PI3K) but does not require activation of the PI3K-Akt pathway. We observed clustering of alpha6beta4 in breast carcinoma cells after adhesion-independent cross-linking of the beta4 integrin subunit. Clustering was significantly blocked when cross-linking was performed in the presence of PI3K inhibitors LY294002 and wortmannin. In contrast, no significant inhibition of clustering was observed with protein kinase C inhibitor GF109203X, rapamycin, or heparin. Although alpha6beta4 clustering was blocked by PI3K inhibitors, clustering was not associated with increased PI3K lipid kinase activity or increased phosphorylation of Akt. A novel role for PI3K in alpha6beta4 integrin clustering is proposed.
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone 3E1, ascites fluid, clone 3E1, Chemicon®