Merck
  • Peripubertal exposure to the neonicotinoid pesticide dinotefuran affects dopaminergic neurons and causes hyperactivity in male mice.
Peripubertal exposure to the neonicotinoid pesticide dinotefuran affects dopaminergic neurons and causes hyperactivity in male mice.
The Journal of veterinary medical science (2018-02-13)
Naoki Yoneda, Tadashi Takada, Tetsushi Hirano, Shogo Yanai, Anzu Yamamoto, Youhei Mantani, Toshifumi Yokoyama, Hiroshi Kitagawa, Yoshiaki Tabuchi, Nobuhiko Hoshi
ABSTRACT
Although neonicotinoid pesticides are expected to have harmful influence on mammals, there is little animal experimental data to support the effect and mechanisms. Since acetylcholine causes the release of dopamine, neonicotinoids may confer a risk of developmental disorders via a disturbance in the monoamine systems. Male mice were peripubertally administered dinotefuran (DIN) referring to no observed effect level (NOEL) and performed behavioral and immunohistological analyses. In an open field test, the total locomotor activity was increased in a dose-dependent manner. The immunoreactivity of tyrosine hydroxylase in the substantia nigra was increased in DIN-exposed mice. These results suggest that exposure to DIN in peripubertal male mice causes hyperactivity and a disturbance of dopaminergic signaling.
Sigma-Aldrich
Anti-Dopamine D2 Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Dopamine D1A Receptor Antibody, clone SG2-D1a, clone SG2-D1a, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®