Merck
  • Human chorionic gonadotropin promotes recruitment of regulatory T cells in endometrium by inducing chemokine CCL2.
Human chorionic gonadotropin promotes recruitment of regulatory T cells in endometrium by inducing chemokine CCL2.
Journal of reproductive immunology (2019-12-07)
Xiaomin Huang, Yunni Cai, Min Ding, Bo Zheng, Haixiang Sun, Jianjun Zhou
ABSTRACT
Human chorionic gonadotropin (hCG) can attract regulatory T cells (Tregs) into the fetal-maternal interface regulating maternal immune tolerance in pregnancy. The objective of this study was to investigate whether hCG recruits the Tregs into endometrium by inducing chemokines. The number of Tregs in the endometrium was analyzed by immunohistochemistry. The expression of CCL2 was analyzed in vivo and in vitro with hCG stimulation. CD4+CD25+ Tregs were isolated from peripheral blood for Tregs migration assay with hCG, CCL2 siRNA and CCR2 antagonist stimulation. Our results showed that the number of endometrial Tregs in RIF patients was significantly decreased (9.4 ± 5.3 vs. 23.1 ± 3.1, P < 0.01), while intrauterine infusion of 2000 IU hCG increased the endometrial Tregs (18.6 ± 9.8 vs. 9.4 ± 5.3, P < 0.05) and CCL2 expression (0.21 ± 0.01 vs. 0.17 ± 0.01, P < 0.01). Horn injecting with 10 IU hCG also increased the endometrial Tregs in pseudopregnant mice (46 ± 16.8 vs. 7 ± 4.3, P < 0.01). Furthermore, the CCL2 protein and mRNA levels were significantly increased in human endometrial stromal cells (hESCs) with the stimulation of hCG. Migration assays showed that hESCs with hCG stimulation promoted Tregs migration (2597 ± 833.2 vs. 1115 ± 670.7, P < 0.05), while the number of migrated Tregs significantly decreased with CCL2 siRNA (1061 ± 105.4 vs. 2598 ± 294.7, P < 0.05) or CCR2 antagonist (356.7 ± 138.8 vs. 2597 ± 833.2, P < 0.05) treatment. In conclusion, intrauterine perfusion of hCG might promote the recruitment of Tregs into endometrium by inducing chemokine CCL2.