Merck
  • Despite Predominance of Uropathogenic/Extraintestinal Pathotypes Among Travel-acquired Extended-spectrum β-Lactamase-producing Escherichia coli, the Most Commonly Associated Clinical Manifestation Is Travelers' Diarrhea.
Despite Predominance of Uropathogenic/Extraintestinal Pathotypes Among Travel-acquired Extended-spectrum β-Lactamase-producing Escherichia coli, the Most Commonly Associated Clinical Manifestation Is Travelers' Diarrhea.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2019-04-30)
Anu Kantele, Tinja Lääveri, Sointu Mero, Inka M K Häkkinen, Juha Kirveskari, Brian D Johnston, James R Johnson
ABSTRACT
One-third of the 100 million travelers to the tropics annually acquire extended-spectrum β-lactamase (ESBL)-producing Enterobacteriaceae (ESBL-PE), with undefined clinical consequences. Symptoms suggesting Enterobacteriaceae infections were recorded prospectively among 430 Finnish travelers, 90 (21%) of whom acquired ESBL-PE abroad. ESBL-PE isolates underwent polymerase chain reaction-based detection of diarrheagenic Escherichia coli (DEC) pathotypes (enteroaggregative E. coli [EAEC], enteropathogenic E. coli [EPEC], enterotoxigenic E. coli [ETEC], enteroinvasive E. coli, and Shiga toxin-producing E. coli), and extraintestinal pathogenic/uropathogenic E. coli (ExPEC/UPEC). Laboratory-confirmed ESBL-PE infections were surveyed 5 years before and after travel. Among the 90 ESBL-PE carriers, manifestations of Enterobacteriaceae infection included travelers' diarrhea (TD) (75/90 subjects) and urinary tract infection (UTI) (3/90). The carriers had 96 ESBL-producing E. coli isolates, 51% exhibiting a molecular pathotype: 13 (14%) were DEC (10 EAEC, 2 EPEC, 1 ETEC) (12 associated with TD) and 39 (41%) ExPEC/UPEC (none associated with UTI). Of ESBL-PE, 3 (3%) were ExPEC/UPEC-EAEC hybrids (2 associated with diarrhea, none with UTI). Potential ESBL-PE infections were detected in 15 of 90 subjects (17%). The 10-year medical record survey identified 4 laboratory-confirmed ESBL-PE infections among the 430 travelers, all in subjects who screened ESBL-PE negative after returning home from their index journeys but had traveled abroad before their infection episodes. Half of all travel-acquired ESBL-producing E. coli strains qualified molecularly as pathogens. Extraintestinal and uropathogenic pathotypes outnumbered enteric pathotypes (41% vs 14%), yet the latter correlated more closely with symptomatic infection (0% vs 92%). Despite more ESBL-PE strains qualifying as ExPEC/UPEC than DEC, travel-acquired ESBL-PE are more often associated with TD than UTI.