Merck
Loss of Setd4 delays radiation-induced thymic lymphoma in mice.
DNA repair (2019-12-04)
Xing Feng, Huimei Lu, Jingyin Yue, Neta Schneider, Jingmei Liu, Lisa K Denzin, Chang S Chan, Subhajyoti De, Zhiyuan Shen
ABSTRACT
Radiation-induced lymphomagenesis results from a clonogenic lymphoid cell proliferation due to genetic alterations and immunological dysregulation. Mouse models had been successfully used to identify risk and protective factors for radiation-induced DNA damage and carcinogenesis. The mammalian SETD4 is a poorly understood putative methyl-transferase. Here, we report that conditional Setd4 deletion in adult mice significantly extended the survival of radiation-induced T-lymphoma. However, in Tp53 deficient mice, Setd4 deletion did not delay the radiation-induced lymphomagenesis although it accelerated the spontaneous T-lymphomagenesis in non-irradiated mice. The T-lymphomas were largely clonogenic in both Setd4flox/flox and Setd4Δ/Δ mice based on sequencing analysis of the T-cell antigen β receptors. However, the Setd4Δ/Δ T-lymphomas were CD4+/CD8+ double positive, while the littermate Setd4flox/floxtumor were largely CD8+ single positive. A genomic sequencing analysis on chromosome deletion, inversion, duplication, and translocation, revealed a larger contribution of inversion but a less contribution of deletion to the overall chromosome rearrangements in the in Setd4Δ/Δ tumors than the Setd4flox/flox tumors. In addition, the Setd4flox/flox mice died more often from the large sizes of primary thymus lymphoma at earlier time, but there was a slight increase of lymphoma dissemination among peripheral organs in Setd4Δ/Δ at later times. These results suggest that Setd4 has a critical role in modulating lymphomagenesis and may be targeted to suppress radiation-induced carcinogenesis.
Sigma-Aldrich
Tamoxifen, ≥99%