Merck
  • Differential investment in pre- and post-mating male sexual traits in response to prolonged exposure to ambient UVB radiation in a fish.
Differential investment in pre- and post-mating male sexual traits in response to prolonged exposure to ambient UVB radiation in a fish.
The Science of the total environment (2020-01-14)
Simon Vitt, Theo C M Bakker, Ingolf P Rick
ABSTRACT
Increasing UVB radiation (UVB) reaching earth's surface following stratospheric ozone depletion is linked to serious consequences for organisms. While studies have focused on direct cytocidal and immunomodulatory effects of UVB, indirect consequences for fitness-related life-history traits are largely unexplored, although knowledge is needed to understand organismal responses to climate change. The present study investigates the effects of developmental exposure to enhanced, but ecologically relevant, ambient UVB levels on growth (body size), parental behavior (nest-building), fitness-enhancing traits of pre-mating (sexual ornamentation) and post-mating (sperm traits) sexual selection as well as their interrelationships in male three-spined sticklebacks (Gasterosteus aculeatus). Moreover, potentially underlying direct UVB effects, testicular antioxidant capacity (TAC) and testes and skin melanization were quantified. Juvenile full-siblings were split into two groups and raised until adulthood in a semi-natural set-up under light conditions including either natural (UVBnormal) or elevated UVB levels (UVBenhanced). When becoming reproductive, males were kept individually before data collection took place at their reproductive peak. The results showed that males from the UVBenhanced-group were smaller than their brothers from the UVBnormal-treatment, whereas no treatment differences were observed for sexual ornamentation, sperm traits and TAC. Moreover, UVB-stressed males built their nests faster and the relationship between body size and nest size was negative for UVBenhanced-males while being positive for UVBnormal-males. These results demonstrate that, depending on physical state, UVB-stressed males adjusted their behavior to some extent. Additionally, a trade-off between the investment in pre- and post-mating traits was found for UVBenhanced-males, i.e. the intensity of their breeding coloration was negatively correlated with sperm number whereas this relationship was reversed for UVBnormal-males, thus showing an interaction between pre- and post-mating traits regarding exposure treatment. The interaction provides first experimental evidence that differential allocation to energetically demanding pre- and post-mating components of male fitness is triggered by a key environmental stressor of climate change.
Sigma-Aldrich
Total Antioxidant Capacity Assay Kit, Sufficient for 100 Colorimetric tests