Merck
  • Amperometric detection of Enterobacteriaceae in river water by measuring β-galactosidase activity at interdigitated microelectrode arrays.
Amperometric detection of Enterobacteriaceae in river water by measuring β-galactosidase activity at interdigitated microelectrode arrays.
Analytica chimica acta (2010-09-15)
Olivier Laczka, Cristina García-Aljaro, Francisco Javier del Campo, Francesc Xavier Muñoz Pascual, Jordi Mas-Gordi, Eva Baldrich
ABSTRACT
Two simple methodologies are compared for the detection of faecal contamination in water using amperometry at gold interdigitated microelectrodes. They rely on the detection of β-galactosidase (β-gal) by redox cycling amperometry of the p-aminophenol (PAP) produced by the enzyme from the 4-aminophenyl β-d-galactopyranoside (PAPG) substrate. The use of phages as specific agents for the release of the bacteria-enclosed enzyme allowed the detection of 6×10(5) CFU mL(-1)Escherichia coli in 2 h without any pre-enrichment or preconcentration steps. Better limits of detection were achieved for the second strategy in the absence of phages. In this case, bacteria were enriched in the presence of both β-d-1-thiogalactopyranoside (IPTG) and substrate but in the absence of phages. Under such experimental conditions, 5×10(4) CFU mL(-1) E. coli could be detected after 2 h of incubation, while 7 h of incubation were enough to detect down to 10 CFU mL(-1) in river water samples. This represents a straightforward one-step method for the detection of faecal contamination that can be conducted in a single working day with minimal sample manipulation by the user.