Merck
  • Cell surface annexins regulate ADAM-mediated ectodomain shedding of proamphiregulin.
Cell surface annexins regulate ADAM-mediated ectodomain shedding of proamphiregulin.
Molecular biology of the cell (2012-03-23)
Hironao Nakayama, Shinji Fukuda, Hirofumi Inoue, Hisayo Nishida-Fukuda, Yuji Shirakata, Koji Hashimoto, Shigeki Higashiyama
ABSTRACT
A disintegrin and metalloproteinase (ADAM) is a family of enzymes involved in ectodomain shedding of various membrane proteins. However, the molecular mechanism underlying substrate recognition by ADAMs remains unknown. In this study, we successfully captured and analyzed cell surface transient assemblies between the transmembrane amphiregulin precursor (proAREG) and ADAM17 during an early shedding phase, which enabled the identification of cell surface annexins as components of their shedding complex. Annexin family members annexin A2 (ANXA2), A8, and A9 interacted with proAREG and ADAM17 on the cell surface. Shedding of proAREG was increased when ANXA2 was knocked down but decreased with ANXA8 and A9 knockdown, because of enhanced and impaired association with ADAM17, respectively. Knockdown of ANXA2 and A8 in primary keratinocytes altered wound-induced cell migration and ultraviolet B-induced phosphorylation of epidermal growth factor receptor (EGFR), suggesting that annexins play an essential role in the ADAM-mediated ectodomain shedding of EGFR ligands. On the basis of these data, we propose that annexins on the cell surface function as "shedding platform" proteins to determine the substrate selectivity of ADAM17, with possible therapeutic potential in ADAM-related diseases.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Pricing and availability is not currently available.