Merck
  • Functional characterization of a WRKY family gene involved in somatic embryogenesis in Panax ginseng.
Functional characterization of a WRKY family gene involved in somatic embryogenesis in Panax ginseng.
Protoplasma (2019-11-25)
Yu Yang, Ni Wang, Shoujing Zhao
ABSTRACT
As a perennial herbaceous species, Panax ginseng is widely cultivated and used as traditional herbal medicine. The root of Panax ginseng commonly remains expensive as conventional breeding of Panax ginseng is difficult. Somatic embryogenesis (S.E.) is a useful tool for plant propagation and optimal model for understanding the mechanisms of plant embryogenesis. In Panax ginseng, increasing studies have been widely performed to optimize the technology of S.E., while the underlying mechanism remains unclear. In this paper, we cloned and identified a WRKY family gene named PgWRKY6 which is upregulated in response to 2,4-D (2,4-dichlorophenoxyacetic acid)-induced embryogenic callus development. The silencing of PgWRKY6 obviously reduces the induction rate of embryogenic callus, indicating its crucial role in S.E. of Panax ginseng hairy root. The expressions of several ROS-scavenging genes are also inducible during embryogenic callus development, and the transcriptions of PgGST, PgAPX1, and PgSOD are demonstrated to be regulated by PgWRKY6. Recombinant PgWRKY6, an approximate 40-KDa protein purified from Escherichia coli, shows a specific DNA-binding activity with a potential recognition site of TTGAC(C/T). This work demonstrated that as a conserved WRKY family transcription factor, PgWRKY6 functions upstream of PgGST, PgAPX1, and PgSOD, and potentially mediated auxins -ROS signaling pathway in the process of S.E. in Panax species.