Merck
  • Simultaneous quantification of different cyclodextrins and Gantrez by HPLC with evaporative light scattering detection.
Simultaneous quantification of different cyclodextrins and Gantrez by HPLC with evaporative light scattering detection.
Journal of pharmaceutical and biomedical analysis (2005-06-11)
M Agüeros, M A Campanero, J M lrache
ABSTRACT
A rapid and simple HPLC method with evaporative scattering detection (ELSD) has been developed for the separation and quantitation of beta-cyclodextrin (beta-CD), 2-hydroxypropyl-beta-cyclodextrin (2-HP-beta-CD) and poly(methyl vinyl ether-co-maleic anhydride) (Gantrez). Separation was carried out on a Zorbax Eclipse XDB-Phenyl narrow bore column, with water-acetonitrile in gradient elution as mobile phase at a flow-rate of 0.25 ml/min. Polyethylenglycol 6000 was used as internal standard. The limit of quantification was of about 0.2 mg/ml for cyclodextrins and 0.05 mg/ml for Gantrez. The precision did not exceed 7%. This method was successfully applied to the rapid analysis of CD-Gantrez nanoparticle conjugates without interference from other components of the formulation.