Merck
  • Genetic variants of N-acetyltransferases 1 and 2 (NAT1 and NAT2) in cynomolgus and rhesus macaques.
Genetic variants of N-acetyltransferases 1 and 2 (NAT1 and NAT2) in cynomolgus and rhesus macaques.
Biochemical pharmacology (2020-04-28)
Yasuhiro Uno, Norie Murayama, Hiroshi Yamazaki
ABSTRACT
In humans, polymorphic N-acetyltransferases NAT1 and NAT2 are important enzymes that metabolize endogenous and exogenous compounds, including drugs. These enzymes exhibit considerable inter-individual variability in humans. The cynomolgus macaque is a nonhuman primate species that is widely used in drug metabolism studies. NAT1/2 in these macaques have molecular and enzymatic similarities to their human orthologs; however, genetic polymorphisms in NAT1/2 have not been fully investigated in this species. In this study, the resequencing of NAT1 and NAT2 in 114 cynomolgus macaques and 19 rhesus macaques found 15 non-synonymous variants for NAT1 and 11 non-synonymous variants and 1 insertion/deletion variant for NAT2. Nine (60%) and five (33%) NAT1 variants and seven (67%) and three (25%) NAT2 variants were unique to cynomolgus and rhesus macaques, respectively. Functional characterization of the mutant enzymes was carried out using cynomolgus NAT1 and NAT2 proteins heterologously expressed in Escherichia coli. Compared with wild-type NAT1, the D122N NAT1 variant showed substantially lower acetylation activities toward p-aminobenzoic acid but had higher acetylation activities toward isoniazid. Moreover, liver cytosolic fractions from cynomolgus macaques homozygous for T98A NAT2 showed significantly lower acetylation activities toward isoniazid than wild-type NAT2; similar results were obtained for recombinant T98A NAT2. Interestingly, all the rhesus macaques analyzed were homozygous for T98A. These findings indicate that polymorphic NAT1/2 variants in cynomolgus and rhesus macaques, especially the T98A NAT2 variant, could account for the inter-animal and/or inter-lineage variabilities of NAT-dependent drug metabolism in macaques.
Sigma-Aldrich
4-Acetamidobenzoic acid, 98%
Pricing and availability is not currently available.