Merck
  • miR-199a-5p Exacerbated Intestinal Barrier Dysfunction through Inhibiting Surfactant Protein D and Activating NF-κB Pathway in Sepsis.
miR-199a-5p Exacerbated Intestinal Barrier Dysfunction through Inhibiting Surfactant Protein D and Activating NF-κB Pathway in Sepsis.
Mediators of inflammation (2020-06-09)
Xianjin Du, Dan Tian, Jie Wei, Chen Yan, Peng Hu, Xu Wu, Wenbin Yang, Zhanyong Zhu
ABSTRACT
Sepsis is a severe disease, which results from the excessive inflammatory response to the infection. Dysfunction of intestinal barrier is a crucial problem in various pathological conditions. Meanwhile, microRNAs exhibit significant roles in the modulation of many diseases, including sepsis. Multiple investigations indicate that miR-199a-5p participates in different human diseases. Nevertheless, little is known on the roles of miR-199a-5p in sepsis. Herein, we evaluated the mechanism of miR-199a-5p on the intestinal barrier dysfunction in sepsis. Intestinal mucosa permeability indicators including D-lactic acid, DAO, and FD-40 levels were determined, and they were greatly increased in sepsis. Then, we proved that miR-199a-5p was induced in sepsis mice tissues and isolated intestinal epithelial cells. Moreover, miR-199a-5p increased D-lactic acid, DAO, and FD-40 while inhibition of miR-199a-5p exhibited a reversed process. Additionally, we observed that miR-199a-5p affected the oxidative damage and inflammation in the intestine tissues from sepsis mice. The content of MDA was elevated whereas SOD was remarkably repressed in the miR-199a-5p mimic group. IL-6, IL-1β, and TNF-α were induced by miR-199a-5p overexpression while IL-10 was reduced by miR-199a-5p. Subsequently, surfactant protein D (SP-D) was predicted as the target of miR-199a-5p. The activation of NF-κB has been identified in sepsis. Herein, we demonstrated that inhibitor of miR-199a-5p contributed to IEC injury via targeting SP-D and inactivating the NF-κB pathway. These revealed miR-199a-5p exacerbated the intestinal barrier dysfunction via inhibiting SP-D and activating the NF-κB pathway in sepsis.