Merck
  • Traction and attraction: haptotaxis substrates collagen and fibronectin interact with chemotaxis by HGF to regulate myoblast migration in a microfluidic device.
Traction and attraction: haptotaxis substrates collagen and fibronectin interact with chemotaxis by HGF to regulate myoblast migration in a microfluidic device.
American journal of physiology. Cell physiology (2020-04-30)
Ziba Roveimiab, Francis Lin, Judy E Anderson
ABSTRACT
Cell migration is central to development, wound healing, tissue regeneration, and immunity. Despite extensive knowledge of muscle regeneration, myoblast migration during regeneration is not well understood. C2C12 mouse myoblast migration and morphology were investigated using a triple-docking polydimethylsiloxane-based microfluidic device in which cells moved under gravity-driven laminar flow on uniform (=) collagen (CN=), fibronectin (FN=), or opposing gradients (CN-FN or FN-CN). In haptotaxis experiments, migration was faster on FN= than on CN=. At 10 h, cells were more elongated on FN-CN and migration was faster than on the CN-FN substrate. Net migration distance on FN-CN at 10 h was greater than on CN-FN, as cells rapidly entered the channel as a larger population (bulk-cell movement, wave 1). Hepatocyte growth factor (HGF) stimulated rapid chemotaxis on FN= but not CN=, increasing migration speed at 10 h early in the channel at low HGF in a steep HGF gradient. HGF accelerated migration on FN= and bulk-cell movement on both uniform substrates. An HGF gradient also slowed cells in wave 2 moving on FN-CN, not CN-FN. Both opposing-gradient substrates affected the shape, speed, and net distance of migrating cells. Gradient and uniform configurations of HGF and substrate differentially influenced migration behavior. Therefore, haptotaxis substrate configuration potently modifies myoblast chemotaxis by HGF. Innovative microfluidic experiments advance our understanding of intricate complexities of myoblast migration. Findings can be leveraged to engineer muscle-tissue volumes for transplantation after serious injury. New analytical approaches may generate broader insights into cell migration.